Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600597 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLCL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLCL1 - phospholipase C like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006226.3 | 446 | Silent Mutation | GCC,GCG | A,A 16 | NP_006217.3 | |
XM_005246643.3 | 446 | Intron | XP_005246700.1 | |||
XM_005246644.3 | 446 | Intron | XP_005246701.1 | |||
XM_011511351.2 | 446 | UTR 5 | XP_011509653.1 | |||
XM_017004339.1 | 446 | Intron | XP_016859828.1 | |||
XM_017004340.1 | 446 | Intron | XP_016859829.1 |