Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612491 MIM: 608769 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APIP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
APIP - APAF1 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015957.3 | 634 | Intron | NP_057041.2 | |||
XM_011520154.2 | 634 | Intron | XP_011518456.1 | |||
XM_017017875.1 | 634 | Intron | XP_016873364.1 |
PDHX - pyruvate dehydrogenase complex component X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135024.1 | 634 | Intron | NP_001128496.1 | |||
NM_001166158.1 | 634 | Silent Mutation | TCC,TCT | S,S 36 | NP_001159630.1 | |
NM_003477.2 | 634 | Silent Mutation | TCC,TCT | S,S 36 | NP_003468.2 | |
XM_011520390.1 | 634 | Intron | XP_011518692.1 |
Set Membership: |
HapMap |