Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605561 MIM: 605478 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PKP3 - plakophilin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIGIRR - single Ig and TIR domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135053.1 | 1601 | Silent Mutation | GTC,GTT | V,V 362 | NP_001128525.1 | |
NM_001135054.1 | 1601 | Silent Mutation | GTC,GTT | V,V 362 | NP_001128526.1 | |
NM_021805.2 | 1601 | Silent Mutation | GTC,GTT | V,V 362 | NP_068577.2 | |
XM_005253044.2 | 1601 | Missense Mutation | CTT,TTT | L,F 459 | XP_005253101.1 | |
XM_005253045.1 | 1601 | Missense Mutation | CTT,TTT | L,F 459 | XP_005253102.1 | |
XM_005253046.1 | 1601 | Missense Mutation | CTT,TTT | L,F 459 | XP_005253103.1 | |
XM_005253048.4 | 1601 | Silent Mutation | GTC,GTT | V,V 376 | XP_005253105.1 | |
XM_017018099.1 | 1601 | Silent Mutation | GTC,GTT | V,V 431 | XP_016873588.1 | |
XM_017018100.1 | 1601 | Silent Mutation | GTC,GTT | V,V 375 | XP_016873589.1 |