Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603605 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIMP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AIMP1 - aminoacyl tRNA synthetase complex interacting multifunctional protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142415.1 | 2902 | Missense Mutation | CCA,GCA | P,A 79 | NP_001135887.1 | |
NM_001142416.1 | 2902 | Missense Mutation | CCA,GCA | P,A 103 | NP_001135888.1 | |
NM_004757.3 | 2902 | Missense Mutation | CCA,GCA | P,A 79 | NP_004748.2 | |
XM_017008835.1 | 2902 | Missense Mutation | CCA,GCA | P,A 79 | XP_016864324.1 | |
XM_017008836.1 | 2902 | Missense Mutation | CCA,GCA | P,A 79 | XP_016864325.1 |
Set Membership: |
HapMap |