Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFX8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RFX8 - RFX family member 8, lacking RFX DNA binding domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145664.1 | Intron | NP_001139136.1 | ||||
XM_011511771.2 | Intron | XP_011510073.1 | ||||
XM_011511772.1 | Intron | XP_011510074.1 | ||||
XM_011511773.1 | Intron | XP_011510075.1 | ||||
XM_011511776.1 | Intron | XP_011510078.1 | ||||
XM_011511777.1 | Intron | XP_011510079.1 | ||||
XM_011511778.1 | Intron | XP_011510080.1 | ||||
XM_017004851.1 | Intron | XP_016860340.1 | ||||
XM_017004852.1 | Intron | XP_016860341.1 | ||||
XM_017004853.1 | Intron | XP_016860342.1 | ||||
XM_017004854.1 | Intron | XP_016860343.1 |
Set Membership: |
HapMap |