Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RPL31 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPL31 - ribosomal protein L31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000993.4 | 3313 | Intron | NP_000984.1 | |||
NM_001098577.2 | 3313 | Intron | NP_001092047.1 | |||
NM_001099693.1 | 3313 | Intron | NP_001093163.1 |
TBC1D8 - TBC1 domain family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005263862.3 | 3313 | Nonsense Mutation | AAA,TAA | K,* 987 | XP_005263919.1 | |
XM_005263863.4 | 3313 | Missense Mutation | AAA,TAA | K,* 980 | XP_005263920.1 | |
XM_005263864.3 | 3313 | Nonsense Mutation | AAA,TAA | K,* 972 | XP_005263921.1 | |
XM_011510507.1 | 3313 | Nonsense Mutation | AAA,TAA | K,* 959 | XP_011508809.1 | |
XM_011510508.2 | 3313 | Intron | XP_011508810.1 | |||
XM_017003198.1 | 3313 | Nonsense Mutation | AAA,TAA | K,* 952 | XP_016858687.1 | |
XM_017003199.1 | 3313 | Missense Mutation | AAA,TAA | K,* 937 | XP_016858688.1 | |
XM_017003200.1 | 3313 | Intron | XP_016858689.1 | |||
XM_017003201.1 | 3313 | Intron | XP_016858690.1 | |||
XM_017003202.1 | 3313 | Intron | XP_016858691.1 |