Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608112 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRAK1 - trafficking kinesin protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042646.2 | Intron | NP_001036111.1 | ||||
NM_001265608.1 | Intron | NP_001252537.1 | ||||
NM_001265609.1 | Intron | NP_001252538.1 | ||||
NM_001265610.1 | Intron | NP_001252539.1 | ||||
NM_014965.4 | Intron | NP_055780.2 | ||||
XM_005264956.2 | Intron | XP_005265013.1 | ||||
XM_005264958.2 | Intron | XP_005265015.1 | ||||
XM_005264960.3 | Intron | XP_005265017.1 | ||||
XM_005264962.3 | Intron | XP_005265019.1 | ||||
XM_005264963.3 | Intron | XP_005265020.1 | ||||
XM_006713029.2 | Intron | XP_006713092.1 | ||||
XM_006713030.2 | Intron | XP_006713093.1 | ||||
XM_006713031.3 | Intron | XP_006713094.1 | ||||
XM_011533489.2 | Intron | XP_011531791.1 | ||||
XM_017005906.1 | Intron | XP_016861395.1 | ||||
XM_017005907.1 | Intron | XP_016861396.1 | ||||
XM_017005908.1 | Intron | XP_016861397.1 | ||||
XM_017005909.1 | Intron | XP_016861398.1 | ||||
XM_017005910.1 | Intron | XP_016861399.1 | ||||
XM_017005911.1 | Intron | XP_016861400.1 | ||||
XM_017005912.1 | Intron | XP_016861401.1 | ||||
XM_017005913.1 | Intron | XP_016861402.1 |