Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602012 MIM: 602030 MIM: 605798 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf139 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C9orf139 - chromosome 9 open reading frame 139 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ENTPD2 - ectonucleoside triphosphate diphosphohydrolase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FUT7 - fucosyltransferase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPDC1 - neural proliferation, differentiation and control 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015392.3 | 1417 | UTR 3 | NP_056207.3 |