Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616408 MIM: 137020 | ||||||||||||||||||||
Literature Links: |
PYCRL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PYCRL - pyrroline-5-carboxylate reductase-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSTA3 - tissue specific transplantation antigen P35B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317783.1 | 1217 | UTR 3 | NP_001304712.1 | |||
NM_003313.3 | 1217 | UTR 3 | NP_003304.1 | |||
XM_005251051.3 | 1217 | UTR 3 | XP_005251108.2 | |||
XM_011517269.1 | 1217 | UTR 3 | XP_011515571.1 |