Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAAATATGCATTACCATGAAAAC[A/G]TTAAAGAAAAGCAGTCTTAACACTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606400 MIM: 605612 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAPN7 - calpain 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3BP5 - SH3 domain binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018009.3 | 2278 | UTR 3 | NP_001018009.2 | |||
NM_004844.4 | 2278 | UTR 3 | NP_004835.2 | |||
XM_011534251.2 | 2278 | UTR 3 | XP_011532553.1 | |||
XM_011534253.1 | 2278 | UTR 3 | XP_011532555.1 | |||
XM_017007521.1 | 2278 | UTR 3 | XP_016863010.1 | |||
XM_017007522.1 | 2278 | UTR 3 | XP_016863011.1 | |||
XM_017007523.1 | 2278 | UTR 3 | XP_016863012.1 | |||
XM_017007524.1 | 2278 | UTR 3 | XP_016863013.1 | |||
XM_017007525.1 | 2278 | UTR 3 | XP_016863014.1 |
SH3BP5-AS1 - SH3BP5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |