Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600167 | ||||||||||||||||||||
Literature Links: |
HRH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HRH1 - histamine receptor H1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000861.3 | Intron | NP_000852.1 | ||||
NM_001098211.1 | Intron | NP_001091681.1 | ||||
NM_001098212.1 | Intron | NP_001091682.1 | ||||
NM_001098213.1 | Intron | NP_001091683.1 | ||||
XM_011533652.1 | Intron | XP_011531954.1 | ||||
XM_011533653.2 | Intron | XP_011531955.1 | ||||
XM_017006283.1 | Intron | XP_016861772.1 | ||||
XM_017006284.1 | Intron | XP_016861773.1 |