Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609632 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDPD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GDPD5 - glycerophosphodiester phosphodiesterase domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030792.6 | 3258 | UTR 3 | NP_110419.5 | |||
XM_006718697.3 | 3258 | UTR 3 | XP_006718760.1 | |||
XM_011545276.2 | 3258 | UTR 3 | XP_011543578.1 | |||
XM_011545277.2 | 3258 | UTR 3 | XP_011543579.1 | |||
XM_011545278.2 | 3258 | UTR 3 | XP_011543580.1 | |||
XM_011545279.2 | 3258 | UTR 3 | XP_011543581.1 | |||
XM_011545280.1 | 3258 | UTR 3 | XP_011543582.1 | |||
XM_011545281.2 | 3258 | UTR 3 | XP_011543583.1 | |||
XM_011545285.2 | 3258 | UTR 3 | XP_011543587.1 | |||
XM_011545286.1 | 3258 | UTR 3 | XP_011543588.1 | |||
XM_011545287.1 | 3258 | UTR 3 | XP_011543589.1 |
KLHL35 - kelch like family member 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039548.2 | 3258 | Intron | NP_001034637.2 | |||
XM_011544954.2 | 3258 | Intron | XP_011543256.1 |
Set Membership: |
HapMap |