Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606867 MIM: 612167 | ||||||||||||||||||||
Literature Links: |
GORASP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GORASP1 - golgi reassembly stacking protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278789.1 | 3628 | UTR 3 | NP_001265718.1 | |||
NM_001278790.1 | 3628 | UTR 3 | NP_001265719.1 | |||
NM_031899.3 | 3628 | UTR 3 | NP_114105.1 | |||
XM_006713301.3 | 3628 | UTR 3 | XP_006713364.1 | |||
XM_011534020.2 | 3628 | UTR 3 | XP_011532322.1 | |||
XM_011534021.2 | 3628 | UTR 3 | XP_011532323.1 | |||
XM_017007050.1 | 3628 | UTR 3 | XP_016862539.1 | |||
XM_017007051.1 | 3628 | UTR 3 | XP_016862540.1 | |||
XM_017007052.1 | 3628 | UTR 3 | XP_016862541.1 |
WDR48 - WD repeat domain 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303402.1 | 3628 | Intron | NP_001290331.1 | |||
NM_001303403.1 | 3628 | Intron | NP_001290332.1 | |||
NM_020839.3 | 3628 | Intron | NP_065890.1 | |||
XM_005265346.3 | 3628 | Intron | XP_005265403.1 | |||
XM_011533971.1 | 3628 | Intron | XP_011532273.1 | |||
XM_017006960.1 | 3628 | Intron | XP_016862449.1 |