Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604015 MIM: 602380 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
B4GALT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
B4GALT4 - beta-1,4-galactosyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003778.3 | 1966 | UTR 3 | NP_003769.1 | |||
NM_212543.1 | 1966 | UTR 3 | NP_997708.1 | |||
XM_005247855.1 | 1966 | UTR 3 | XP_005247912.1 | |||
XM_006713798.2 | 1966 | UTR 3 | XP_006713861.1 | |||
XM_006713799.2 | 1966 | UTR 3 | XP_006713862.1 | |||
XM_006713800.1 | 1966 | UTR 3 | XP_006713863.1 | |||
XM_006713801.2 | 1966 | UTR 3 | XP_006713864.1 | |||
XM_011513260.1 | 1966 | UTR 3 | XP_011511562.1 | |||
XM_017007401.1 | 1966 | UTR 3 | XP_016862890.1 |
UPK1B - uroplakin 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |