Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600502 MIM: 611834 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGHMBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGHMBP2 - immunoglobulin mu binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002180.2 | 159 | Silent Mutation | CTC,CTT | L,L 19 | NP_002171.2 | |
XM_005273974.2 | 159 | UTR 5 | XP_005274031.1 | |||
XM_005273975.3 | 159 | Intron | XP_005274032.1 | |||
XM_005273976.1 | 159 | Silent Mutation | CTC,CTT | L,L 19 | XP_005274033.1 | |
XM_011544994.1 | 159 | Intron | XP_011543296.1 | |||
XM_017017669.1 | 159 | UTR 5 | XP_016873158.1 | |||
XM_017017670.1 | 159 | Intron | XP_016873159.1 | |||
XM_017017671.1 | 159 | Silent Mutation | CTC,CTT | L,L 19 | XP_016873160.1 |
MRPL21 - mitochondrial ribosomal protein L21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181514.1 | 159 | Intron | NP_852615.1 | |||
NM_181515.1 | 159 | Intron | NP_852616.1 | |||
XM_005273823.4 | 159 | Intron | XP_005273880.1 |