Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHBDD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RHBDD1 - rhomboid domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167608.1 | Intron | NP_001161080.1 | ||||
NM_032276.3 | Intron | NP_115652.2 | ||||
XM_017005083.1 | Intron | XP_016860572.1 | ||||
XM_017005084.1 | Intron | XP_016860573.1 | ||||
XM_017005085.1 | Intron | XP_016860574.1 | ||||
XM_017005086.1 | Intron | XP_016860575.1 | ||||
XM_017005087.1 | Intron | XP_016860576.1 | ||||
XM_017005088.1 | Intron | XP_016860577.1 | ||||
XM_017005089.1 | Intron | XP_016860578.1 | ||||
XM_017005090.1 | Intron | XP_016860579.1 | ||||
XM_017005091.1 | Intron | XP_016860580.1 | ||||
XM_017005092.1 | Intron | XP_016860581.1 | ||||
XM_017005093.1 | Intron | XP_016860582.1 | ||||
XM_017005094.1 | Intron | XP_016860583.1 | ||||
XM_017005095.1 | Intron | XP_016860584.1 |
Set Membership: |
HapMap Validated |