Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615691 MIM: 615320 MIM: 606991 MIM: 614472 | |||||||||||||||||||||||
Literature Links: |
AMIGO3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
AMIGO3 - adhesion molecule with Ig like domain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GMPPB - GDP-mannose pyrophosphorylase B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IP6K1 - inositol hexakisphosphate kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006115.2 | 4014 | UTR 3 | NP_001006115.1 | |||
NM_001242829.1 | 4014 | UTR 3 | NP_001229758.1 | |||
NM_153273.3 | 4014 | UTR 3 | NP_695005.1 |
RNF123 - ring finger protein 123 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |