Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609198 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTSL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADAMTSL1 - ADAMTS like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040272.5 | Intron | NP_001035362.3 | ||||
NM_052866.4 | Intron | NP_443098.3 | ||||
XM_011518063.2 | Intron | XP_011516365.1 | ||||
XM_011518064.2 | Intron | XP_011516366.1 | ||||
XM_011518067.1 | Intron | XP_011516369.1 | ||||
XM_011518068.2 | Intron | XP_011516370.1 | ||||
XM_011518070.2 | Intron | XP_011516372.1 | ||||
XM_017015310.1 | Intron | XP_016870799.1 | ||||
XM_017015311.1 | Intron | XP_016870800.1 | ||||
XM_017015312.1 | Intron | XP_016870801.1 | ||||
XM_017015313.1 | Intron | XP_016870802.1 | ||||
XM_017015314.1 | Intron | XP_016870803.1 |
Set Membership: |
HapMap |