Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAGTCACCTTCGCAACCTGGGA[A/C]GCCTTCTTGCATAAGATGCCTGAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604264 MIM: 606671 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CELSR3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CELSR3 - cadherin EGF LAG seven-pass G-type receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CELSR3-AS1 - CELSR3 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCKIPSD - NCK interacting protein with SH3 domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016453.3 | 2133 | Intron | NP_057537.1 | |||
NM_184231.2 | 2133 | Intron | NP_909119.1 | |||
XM_017006594.1 | 2133 | Missense Mutation | GTC,TTC | V,F 675 | XP_016862083.1 | |
XM_017006595.1 | 2133 | Missense Mutation | GTC,TTC | V,F 668 | XP_016862084.1 | |
XM_017006596.1 | 2133 | Missense Mutation | GTC,TTC | V,F 597 | XP_016862085.1 | |
XM_017006597.1 | 2133 | Missense Mutation | GTC,TTC | V,F 597 | XP_016862086.1 | |
XM_017006598.1 | 2133 | Missense Mutation | GTC,TTC | V,F 590 | XP_016862087.1 | |
XM_017006599.1 | 2133 | Intron | XP_016862088.1 | |||
XM_017006600.1 | 2133 | Intron | XP_016862089.1 |
Set Membership: |
HapMap |