Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 616292 | |||||||||||||||||||||||||||||
Literature Links: |
SAXO1 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SAXO1 - stabilizer of axonemal microtubules 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287049.1 | 1804 | UTR 3 | NP_001273978.1 | |||
NM_001287050.1 | 1804 | UTR 3 | NP_001273979.1 | |||
NM_153707.3 | 1804 | UTR 3 | NP_714918.2 | |||
XM_011517744.1 | 1804 | Intron | XP_011516046.1 | |||
XM_011517745.1 | 1804 | Intron | XP_011516047.1 | |||
XM_011517746.2 | 1804 | Intron | XP_011516048.1 | |||
XM_011517747.2 | 1804 | Intron | XP_011516049.1 | |||
XM_017014314.1 | 1804 | Intron | XP_016869803.1 | |||
XM_017014315.1 | 1804 | Intron | XP_016869804.1 |