Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609471 MIM: 612191 MIM: 108961 MIM: 615742 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GBA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GBA2 - glucosylceramidase beta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSMP - microseminoprotein, prostate associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001044264.2 | Intron | NP_001037729.1 |
NPR2 - natriuretic peptide receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003995.3 | Intron | NP_003986.2 | ||||
XM_005251478.3 | Intron | XP_005251535.1 | ||||
XM_005251479.4 | Intron | XP_005251536.1 | ||||
XM_011517890.2 | Intron | XP_011516192.1 | ||||
XM_011517891.2 | Intron | XP_011516193.1 | ||||
XM_011517892.2 | Intron | XP_011516194.1 | ||||
XM_011517893.2 | Intron | XP_011516195.1 | ||||
XM_011517895.2 | Intron | XP_011516197.1 | ||||
XM_017014745.1 | Intron | XP_016870234.1 | ||||
XM_017014746.1 | Intron | XP_016870235.1 | ||||
XM_017014747.1 | Intron | XP_016870236.1 | ||||
XM_017014748.1 | Intron | XP_016870237.1 | ||||
XM_017014749.1 | Intron | XP_016870238.1 | ||||
XM_017014750.1 | Intron | XP_016870239.1 |
RGP1 - RGP1 homolog, RAB6A GEF complex partner 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080496.2 | Intron | NP_001073965.2 |
Set Membership: |
HapMap |