Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611532 | ||||||||||||||||||||
Literature Links: |
MIR6851 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6851 - microRNA 6851 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOL6 - nucleolar protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022917.4 | 4021 | UTR 3 | NP_075068.2 | |||
NM_139235.3 | 4021 | UTR 3 | NP_631981.2 | |||
XM_017015044.1 | 4021 | UTR 3 | XP_016870533.1 |