Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602867 MIM: 180661 | ||||||||||||||||||||
Literature Links: |
IGFBP7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGFBP7 - insulin like growth factor binding protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253835.1 | 762 | Missense Mutation | GCT,GTT | A,V 243 | NP_001240764.1 | |
NM_001553.2 | 762 | Missense Mutation | GCT,GTT | A,V 243 | NP_001544.1 |
POLR2B - RNA polymerase II subunit B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |