Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604911 | ||||||||||||||||||||
Literature Links: |
CNOT4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNOT4 - CCR4-NOT transcription complex subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008225.2 | 3291 | Intron | NP_001008226.1 | |||
NM_001190847.1 | 3291 | UTR 3 | NP_001177776.1 | |||
NM_001190848.1 | 3291 | Intron | NP_001177777.1 | |||
NM_001190849.1 | 3291 | UTR 3 | NP_001177778.1 | |||
NM_001190850.1 | 3291 | UTR 3 | NP_001177779.1 | |||
NM_013316.3 | 3291 | UTR 3 | NP_037448.2 | |||
XM_017012235.1 | 3291 | UTR 3 | XP_016867724.1 |