Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAGCCCTATTGGAAGAGTGGGGA[C/T]GAGGTGGGAGAAAGCAACCACAACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600542 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928438 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC101928438 - uncharacterized LOC101928438 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR4A3 - nuclear receptor subfamily 4 group A member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006981.3 | 656 | Intron | NP_008912.2 | |||
NM_173199.2 | 656 | Intron | NP_775291.1 | |||
NM_173200.2 | 656 | Intron | NP_775292.1 | |||
XM_017015162.1 | 656 | UTR 5 | XP_016870651.1 |
Set Membership: |
HapMap Validated |