Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605202 MIM: 606556 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NANS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NANS - N-acetylneuraminate synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM14 - tripartite motif containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014788.3 | 1576 | UTR 3 | NP_055603.2 | |||
NM_033219.2 | 1576 | Intron | NP_150088.1 | |||
XM_005252320.3 | 1576 | Intron | XP_005252377.1 | |||
XM_006717338.1 | 1576 | UTR 3 | XP_006717401.1 | |||
XM_011519244.2 | 1576 | UTR 3 | XP_011517546.1 | |||
XM_017015352.1 | 1576 | Intron | XP_016870841.1 | |||
XM_017015353.1 | 1576 | Intron | XP_016870842.1 | |||
XM_017015354.1 | 1576 | Intron | XP_016870843.1 |
Set Membership: |
HapMap |