Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606105 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC44A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC44A1 - solute carrier family 44 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286730.1 | Intron | NP_001273659.1 | ||||
NM_080546.4 | Intron | NP_536856.2 | ||||
XM_005251855.4 | Intron | XP_005251912.1 | ||||
XM_006717027.3 | Intron | XP_006717090.1 | ||||
XM_006717028.3 | Intron | XP_006717091.1 | ||||
XM_006717029.2 | Intron | XP_006717092.1 | ||||
XM_017014559.1 | Intron | XP_016870048.1 | ||||
XM_017014560.1 | Intron | XP_016870049.1 |
Set Membership: |
HapMap |