Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGACAGCAGGCGGCCAGGTGCTCA[A/G]AGGGCACCAAGGTGCCACAACTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C9orf69 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C9orf69 - chromosome 9 open reading frame 69 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256526.1 | 2710 | UTR 3 | NP_001243455.1 | |||
NM_152833.2 | 2710 | UTR 3 | NP_690046.3 |
LOC105376323 - uncharacterized LOC105376323 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107987142 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |