Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605729 MIM: 138600 MIM: 138610 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKNA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AKNA - AT-hook transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317950.1 | 4809 | UTR 3 | NP_001304879.1 | |||
NM_001317952.1 | 4809 | UTR 3 | NP_001304881.1 | |||
NM_030767.5 | 4809 | UTR 3 | NP_110394.3 | |||
XM_005252244.2 | 4809 | UTR 3 | XP_005252301.1 | |||
XM_005252245.1 | 4809 | UTR 3 | XP_005252302.1 | |||
XM_005252247.4 | 4809 | UTR 3 | XP_005252304.1 | |||
XM_005252248.2 | 4809 | UTR 3 | XP_005252305.1 | |||
XM_006717294.1 | 4809 | UTR 3 | XP_006717357.1 | |||
XM_006717295.2 | 4809 | Intron | XP_006717358.1 | |||
XM_011519063.2 | 4809 | UTR 3 | XP_011517365.1 | |||
XM_011519064.2 | 4809 | Intron | XP_011517366.1 | |||
XM_011519065.2 | 4809 | UTR 3 | XP_011517367.2 | |||
XM_011519066.2 | 4809 | Intron | XP_011517368.1 | |||
XM_017015172.1 | 4809 | UTR 3 | XP_016870661.1 |
ORM1 - orosomucoid 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORM2 - orosomucoid 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |