Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616805 | ||||||||||||||||||||
Literature Links: |
MYCT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYCT1 - myc target 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025107.2 | Intron | NP_079383.2 | ||||
XM_011536149.2 | Intron | XP_011534451.1 | ||||
XM_011536150.2 | Intron | XP_011534452.1 | ||||
XM_011536151.2 | Intron | XP_011534453.1 | ||||
XM_011536152.2 | Intron | XP_011534454.1 | ||||
XM_017011319.1 | Intron | XP_016866808.1 |