Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGATGACACTGCACCCCACCCCCC[A/C]CAACCCACTGAATCCTATCCATCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607352 | ||||||||||||||||||||
Literature Links: |
LOC107984124 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC107984124 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017014154.1 | 587 | Intron | XP_016869643.1 |
RHOBTB2 - Rho related BTB domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160036.1 | 587 | Intron | NP_001153508.1 | |||
NM_001160037.1 | 587 | Intron | NP_001153509.1 | |||
NM_015178.2 | 587 | Intron | NP_055993.2 | |||
XM_017013251.1 | 587 | UTR 5 | XP_016868740.1 |