Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107910 | ||||||||||||||||||||||||||||||||
Literature Links: |
CYP19A1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CYP19A1 - cytochrome P450 family 19 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000103.3 | 1875 | UTR 3 | NP_000094.2 | |||
NM_031226.2 | 1875 | UTR 3 | NP_112503.1 | |||
XM_005254191.1 | 1875 | UTR 3 | XP_005254248.1 |
PIRC66 - piwi-interacting RNA cluster 66 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
DME Validated Inventoried |