Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614568 | ||||||||||||||||||||
Literature Links: |
UNC13C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UNC13C - unc-13 homolog C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005254394.4 | Intron | XP_005254451.1 | ||||
XM_017022220.1 | Intron | XP_016877709.1 | ||||
XM_017022221.1 | Intron | XP_016877710.1 | ||||
XM_017022222.1 | Intron | XP_016877711.1 | ||||
XM_017022223.1 | Intron | XP_016877712.1 | ||||
XM_017022224.1 | Intron | XP_016877713.1 | ||||
XM_017022225.1 | Intron | XP_016877714.1 |