Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603868 MIM: 613262 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB27A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RAB27A - RAB27A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004580.4 | 1136 | UTR 3 | NP_004571.2 | |||
NM_183234.2 | 1136 | UTR 3 | NP_899057.1 | |||
NM_183235.2 | 1136 | UTR 3 | NP_899058.1 | |||
NM_183236.2 | 1136 | UTR 3 | NP_899059.1 | |||
XM_005254576.4 | 1136 | UTR 3 | XP_005254633.1 | |||
XM_011521852.1 | 1136 | UTR 3 | XP_011520154.1 | |||
XM_011521854.1 | 1136 | UTR 3 | XP_011520156.1 | |||
XM_011521855.2 | 1136 | UTR 3 | XP_011520157.1 | |||
XM_011521856.2 | 1136 | UTR 3 | XP_011520158.1 |
RSL24D1 - ribosomal L24 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |