Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601953 MIM: 139150 | ||||||||||||||||||||
Literature Links: |
CCNH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCNH - cyclin H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199189.1 | 2661 | Intron | NP_001186118.1 | |||
NM_001239.3 | 2661 | Intron | NP_001230.1 | |||
XM_005248627.3 | 2661 | Intron | XP_005248684.1 | |||
XM_005248629.4 | 2661 | Intron | XP_005248686.1 | |||
XM_011543706.2 | 2661 | UTR 3 | XP_011542008.1 | |||
XM_017010014.1 | 2661 | UTR 3 | XP_016865503.1 |
RASA1 - RAS p21 protein activator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002890.2 | 2661 | UTR 3 | NP_002881.1 | |||
NM_022650.2 | 2661 | UTR 3 | NP_072179.1 | |||
XM_011543525.1 | 2661 | UTR 3 | XP_011541827.1 | |||
XM_011543527.2 | 2661 | Intron | XP_011541829.1 |