Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609297 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEMA5A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEMA5A - semaphorin 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003966.2 | 6833 | UTR 3 | NP_003957.2 | |||
XM_006714506.2 | 6833 | UTR 3 | XP_006714569.1 | |||
XM_006714507.3 | 6833 | UTR 3 | XP_006714570.1 | |||
XM_011514155.2 | 6833 | UTR 3 | XP_011512457.1 | |||
XM_011514156.1 | 6833 | UTR 3 | XP_011512458.1 | |||
XM_011514157.1 | 6833 | UTR 3 | XP_011512459.1 | |||
XM_011514158.1 | 6833 | UTR 3 | XP_011512460.1 | |||
XM_011514159.2 | 6833 | UTR 3 | XP_011512461.1 | |||
XM_017010016.1 | 6833 | UTR 3 | XP_016865505.1 |
Set Membership: |
HapMap |