Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600610 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABPB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GABPB1 - GA binding protein transcription factor beta subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320910.1 | 2405 | UTR 3 | NP_001307839.1 | |||
NM_001320915.1 | 2405 | UTR 3 | NP_001307844.1 | |||
NM_002041.4 | 2405 | Intron | NP_002032.2 | |||
NM_005254.5 | 2405 | UTR 3 | NP_005245.2 | |||
NM_016654.4 | 2405 | UTR 3 | NP_057738.1 | |||
NM_016655.4 | 2405 | Intron | NP_057739.1 | |||
NM_181427.3 | 2405 | Intron | NP_852092.1 | |||
XM_005254274.3 | 2405 | UTR 3 | XP_005254331.1 | |||
XM_011521426.2 | 2405 | UTR 3 | XP_011519728.1 | |||
XM_017022053.1 | 2405 | UTR 3 | XP_016877542.1 | |||
XM_017022054.1 | 2405 | UTR 3 | XP_016877543.1 |