Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616929 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTERF2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MTERF2 - mitochondrial transcription termination factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033050.2 | 948 | Missense Mutation | ATA,GTA | I,V 198 | NP_001028222.1 | |
NM_025198.4 | 948 | Missense Mutation | ATA,GTA | I,V 198 | NP_079474.2 | |
XM_011538767.2 | 948 | Missense Mutation | ATA,GTA | I,V 198 | XP_011537069.1 | |
XM_017019985.1 | 948 | Missense Mutation | ATA,GTA | I,V 198 | XP_016875474.1 | |
XM_017019986.1 | 948 | Missense Mutation | ATA,GTA | I,V 198 | XP_016875475.1 | |
XM_017019987.1 | 948 | Missense Mutation | ATA,GTA | I,V 198 | XP_016875476.1 |
TMEM263 - transmembrane protein 263 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |