Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601586 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGER4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTGER4 - prostaglandin E receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000958.2 | 4387 | Intron | NP_000949.1 | |||
XM_017009656.1 | 4387 | Intron | XP_016865145.1 | |||
XM_017009657.1 | 4387 | Intron | XP_016865146.1 | |||
XM_017009658.1 | 4387 | Intron | XP_016865147.1 | |||
XM_017009659.1 | 4387 | Intron | XP_016865148.1 |
TTC33 - tetratricopeptide repeat domain 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012382.2 | 4387 | UTR 3 | NP_036514.1 | |||
XM_011514003.2 | 4387 | Intron | XP_011512305.1 |
Set Membership: |
HapMap |