Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615114 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF516 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF516 - zinc finger protein 516 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014643.3 | 7652 | UTR 3 | NP_055458.1 | |||
XM_011526269.1 | 7652 | UTR 3 | XP_011524571.1 | |||
XM_011526270.2 | 7652 | UTR 3 | XP_011524572.1 | |||
XM_011526271.2 | 7652 | UTR 3 | XP_011524573.1 | |||
XM_011526272.2 | 7652 | UTR 3 | XP_011524574.1 | |||
XM_011526273.2 | 7652 | UTR 3 | XP_011524575.1 | |||
XM_011526274.2 | 7652 | UTR 3 | XP_011524576.1 | |||
XM_011526275.2 | 7652 | UTR 3 | XP_011524577.1 | |||
XM_011526276.1 | 7652 | UTR 3 | XP_011524578.1 | |||
XM_011526277.2 | 7652 | Intron | XP_011524579.1 | |||
XM_017026097.1 | 7652 | UTR 3 | XP_016881586.1 |