Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123810 | ||||||||||||||||||||
Literature Links: |
CREB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CREB1 - cAMP responsive element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320793.1 | 181 | UTR 5 | NP_001307722.1 | |||
NM_004379.4 | 181 | UTR 5 | NP_004370.1 | |||
NM_134442.4 | 181 | UTR 5 | NP_604391.1 | |||
XM_011510645.1 | 181 | Intron | XP_011508947.1 | |||
XM_011510646.2 | 181 | UTR 5 | XP_011508948.1 | |||
XM_011510647.2 | 181 | Intron | XP_011508949.1 | |||
XM_011510648.2 | 181 | UTR 5 | XP_011508950.1 | |||
XM_011510649.2 | 181 | UTR 5 | XP_011508951.1 | |||
XM_011510650.2 | 181 | Intron | XP_011508952.1 | |||
XM_011510651.2 | 181 | Intron | XP_011508953.1 | |||
XM_017003399.1 | 181 | UTR 5 | XP_016858888.1 | |||
XM_017003400.1 | 181 | UTR 5 | XP_016858889.1 | |||
XM_017003401.1 | 181 | Intron | XP_016858890.1 |