Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605022 MIM: 610276 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAK2 - p21 (RAC1) activated kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002577.4 | 147 | UTR 5 | NP_002568.2 | |||
XM_011512870.2 | 147 | Intron | XP_011511172.1 | |||
XM_017006501.1 | 147 | UTR 5 | XP_016861990.1 |
PIGX - phosphatidylinositol glycan anchor biosynthesis class X | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |