Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616147 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK15 - cyclin dependent kinase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261435.1 | Intron | NP_001248364.1 | ||||
NM_001261436.1 | Intron | NP_001248365.1 | ||||
NM_139158.2 | Intron | NP_631897.1 | ||||
XM_005246781.3 | Intron | XP_005246838.1 | ||||
XM_005246782.4 | Intron | XP_005246839.1 | ||||
XM_011511650.2 | Intron | XP_011509952.1 | ||||
XM_011511652.1 | Intron | XP_011509954.1 | ||||
XM_011511654.2 | Intron | XP_011509956.1 | ||||
XM_011511655.2 | Intron | XP_011509957.1 | ||||
XM_011511656.2 | Intron | XP_011509958.1 | ||||
XM_011511657.2 | Intron | XP_011509959.1 |