Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300070 | ||||||||||||||||||||
Literature Links: |
FGF13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FGF13 - fibroblast growth factor 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139498.1 | 1965 | UTR 3 | NP_001132970.1 | |||
NM_001139500.1 | 1965 | UTR 3 | NP_001132972.1 | |||
NM_001139501.1 | 1965 | UTR 3 | NP_001132973.1 | |||
NM_001139502.1 | 1965 | UTR 3 | NP_001132974.1 | |||
NM_004114.3 | 1965 | UTR 3 | NP_004105.1 | |||
NM_033642.2 | 1965 | UTR 3 | NP_378668.1 | |||
XM_005262399.1 | 1965 | UTR 3 | XP_005262456.1 |