Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300826 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STAG2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
STAG2 - stromal antigen 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042749.2 | Intron | NP_001036214.1 | ||||
NM_001042750.1 | Intron | NP_001036215.1 | ||||
NM_001042751.1 | Intron | NP_001036216.1 | ||||
NM_001282418.1 | Intron | NP_001269347.1 | ||||
NM_006603.4 | Intron | NP_006594.3 | ||||
XM_005262357.2 | Intron | XP_005262414.1 | ||||
XM_005262358.2 | Intron | XP_005262415.1 | ||||
XM_005262359.3 | Intron | XP_005262416.1 | ||||
XM_005262360.2 | Intron | XP_005262417.1 | ||||
XM_005262361.2 | Intron | XP_005262418.1 | ||||
XM_006724727.1 | Intron | XP_006724790.1 | ||||
XM_006724728.3 | Intron | XP_006724791.1 | ||||
XM_011531253.2 | Intron | XP_011529555.1 | ||||
XM_017029232.1 | Intron | XP_016884721.1 | ||||
XM_017029233.1 | Intron | XP_016884722.1 | ||||
XM_017029234.1 | Intron | XP_016884723.1 | ||||
XM_017029235.1 | Intron | XP_016884724.1 |
Set Membership: |
HapMap |