Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608308 MIM: 609699 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABTB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABTB1 - ankyrin repeat and BTB domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MGLL - monoglyceride lipase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003794.2 | 4031 | UTR 3 | NP_001003794.1 | |||
NM_001256585.1 | 4031 | UTR 3 | NP_001243514.1 | |||
NM_007283.6 | 4031 | UTR 3 | NP_009214.1 | |||
XM_011512377.1 | 4031 | UTR 3 | XP_011510679.1 | |||
XM_011512378.1 | 4031 | UTR 3 | XP_011510680.1 | |||
XM_011512379.2 | 4031 | UTR 3 | XP_011510681.1 | |||
XM_011512382.1 | 4031 | UTR 3 | XP_011510684.1 | |||
XM_011512383.2 | 4031 | UTR 3 | XP_011510685.1 | |||
XM_017005662.1 | 4031 | Intron | XP_016861151.1 | |||
XM_017005663.1 | 4031 | Intron | XP_016861152.1 | |||
XM_017005664.1 | 4031 | UTR 3 | XP_016861153.1 | |||
XM_017005665.1 | 4031 | UTR 3 | XP_016861154.1 | |||
XM_017005666.1 | 4031 | UTR 3 | XP_016861155.1 |
Set Membership: |
HapMap |