Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610803 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC41A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SLC41A3 - solute carrier family 41 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008485.1 | 1868 | Intron | NP_001008485.1 | |||
NM_001008486.1 | 1868 | Intron | NP_001008486.1 | |||
NM_001008487.1 | 1868 | UTR 3 | NP_001008487.1 | |||
NM_001164475.1 | 1868 | UTR 3 | NP_001157947.1 | |||
NM_017836.3 | 1868 | UTR 3 | NP_060306.3 | |||
XM_005247559.1 | 1868 | Intron | XP_005247616.1 | |||
XM_005247562.1 | 1868 | Intron | XP_005247619.1 | |||
XM_005247563.2 | 1868 | Intron | XP_005247620.1 | |||
XM_005247564.1 | 1868 | UTR 3 | XP_005247621.1 | |||
XM_005247565.2 | 1868 | Intron | XP_005247622.1 | |||
XM_006713681.2 | 1868 | Intron | XP_006713744.1 | |||
XM_011512945.1 | 1868 | Intron | XP_011511247.1 | |||
XM_017006705.1 | 1868 | Intron | XP_016862194.1 | |||
XM_017006706.1 | 1868 | UTR 3 | XP_016862195.1 | |||
XM_017006707.1 | 1868 | UTR 3 | XP_016862196.1 | |||
XM_017006708.1 | 1868 | UTR 3 | XP_016862197.1 | |||
XM_017006709.1 | 1868 | UTR 3 | XP_016862198.1 |
Set Membership: |
HapMap Validated |