Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACATGTCTGTGAGAGTGCAGCAA[A/C]GGTCCTCGTCTCACCTGAGAAGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615746 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP100 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CFAP100 - cilia and flagella associated protein 100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105374089 - uncharacterized LOC105374089 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZXDC - ZXD family zinc finger C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040653.3 | Intron | NP_001035743.1 | ||||
NM_025112.4 | Intron | NP_079388.3 | ||||
XM_005247757.3 | Intron | XP_005247814.1 | ||||
XM_006713741.2 | Intron | XP_006713804.1 | ||||
XM_011513119.2 | Intron | XP_011511421.1 |
Set Membership: |
HapMap Validated |