Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613350 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC52A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC52A3 - solute carrier family 52 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033409.3 | 1572 | Silent Mutation | AGC,AGT | S,S 411 | NP_212134.3 | |
XM_005260655.3 | 1572 | Silent Mutation | AGC,AGT | S,S 411 | XP_005260712.1 | |
XM_011529148.1 | 1572 | Silent Mutation | AGC,AGT | S,S 411 | XP_011527450.1 |
Set Membership: |
HapMap |