Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACAAAGGGCGGGCCCGGGGCCT[C/T]GTCGCCGCACCACCTGTATTCAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602730 MIM: 604049 | ||||||||||||||||||||
Literature Links: |
ACVR2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACVR2B - activin A receptor type 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001106.3 | Intron | NP_001097.2 | ||||
XM_005265583.3 | Intron | XP_005265640.1 | ||||
XM_017007514.1 | Intron | XP_016863003.1 | ||||
XM_017007515.1 | Intron | XP_016863004.1 | ||||
XM_017007516.1 | Intron | XP_016863005.1 |
ACVR2B-AS1 - ACVR2B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XYLB - xylulokinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |